Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZMYM3 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298112C
Description
ZMYM3 Polyclonal specifically detects ZMYM3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
ZMYM3 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
DXS6673EZinc finger protein 261KIAA0385XFIM, MYM, zinc finger, MYM-type 3, ZNF198L2, ZNF261zinc finger MYM-type protein 3 | |
Produced in rabbits immunized with E. coli-derived Human ZMYM3 fragment. | |
0.1 mL | |
DNA Repair | |
9203 | |
Store at 4°C in the dark. | |
IgG |
Immunofluorescence | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction