Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZMYM3 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100458F
Description
ZMYM3 Polyclonal antibody specifically detects ZMYM3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
ZMYM3 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
DXS6673EZinc finger protein 261KIAA0385XFIM, MYM, zinc finger, MYM-type 3, ZNF198L2, ZNF261zinc finger MYM-type protein 3 | |
The immunogen this antibody was made to, maps to a region between residues 250-350 of human Zinc Finger Protein 261 using the NP_005087.1 entry Q8IY18 (GeneID 9203). | |
0.1 mL | |
DNA Repair | |
9203 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
FITC | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction