Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZMYM3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584980
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584980 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584980 Supplier Invitrogen™ Supplier No. PA584980
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MDPSDFPSPF DPLTLPEKPL AGDLPVDMEF GEDLLESQTA PTRGWAPPGP SPSSGALDLL DTPAGLEKDP GVLDGATE Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 97%.

This gene is located on the X chromosome and is subject to X inactivation. It is highly conserved in vertebrates and most abundantly expressed in the brain. The encoded protein is a component of histone deacetylase-containing multiprotein complexes that function through modifying chromatin structure to keep genes silent. A chromosomal translocation (X;13) involving this gene is associated with X-linked cognitive disability. Several alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZMYM3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZMYM3
Gene Accession No. Q14202
Gene Alias 9030216B10Rik; AW122925; DXHXS6673E protein; DXS6673E; DXS6673El; KIAA0385; MYM; XFIM; Zfp261; zinc finger MYM-type containing 3; zinc finger MYM-type protein 3; Zinc finger protein 261; zinc finger, MYM-type 3; Zmym3; ZNF198L2; ZNF261
Gene Symbols ZMYM3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZMYM3. Recombinant protein control fragment (Product #RP-108652).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9203
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.