Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZMYM4 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10095000JF549
Description
ZMYM4 Polyclonal antibody specifically detects ZMYM4 in Human samples. It is validated for Western BlotSpecifications
ZMYM4 | |
Polyclonal | |
Western Blot | |
CDIR, cell death inhibiting RNA, KIAA0425DKFZp686L0547, MYM, zinc finger MYM-type protein 4, Zinc finger protein 262DKFZp686B09210, zinc finger, MYM-type 4, ZNF198L3, ZNF262 | |
The immunogen recognized by this antibody maps to a region between residue 850 and 900 of human zinc finger, MYM-type 4 using the numbering given in entry NP_005086.2 (GeneID 9202). | |
0.1 mL | |
Cell Biology | |
9202 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction