Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZMYM4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA577269
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Zinc-finger proteins contain DNA-binding domains characterized by the unique role of zinc and have a wide variety of functions such as transcriptional activation or repression. The protein folding and the DNA binding ability are governed by the coordination of a zinc ion. It has been found to be overexpressed in human lung adenocarcinomas and squamous cell carcinomas, and the overexpression of a fragment of the 3'UTR of the ZMYM4 mRNA termed Cell Death Inhibiting RNA (CDIR) protects HeLa cells from IFN-gamma-induced apoptosis, suggesting that ZMYM4 may play a role in tumorigenesis.
Specifications
ZMYM4 | |
Polyclonal | |
Unconjugated | |
ZMYM4 | |
6330503C17Rik; AI480785; AW493829; CDIR; cell death inhibiting RNA; D630001M21; Kiaa0425; mKIAA0425; MYM; MYM type 4; Zfp262; zinc finger MYM-type containing 4; zinc finger MYM-type protein 4; Zinc finger protein 262; zinc finger, MYM-type 4; Zmym4; ZNF198L3; ZNF262 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
67785, 9202 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
A2A791, Q5VZL5 | |
ZMYM4 | |
Synthetic peptide corresponding to amino acids 804-856 of Human ZMYM4. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction