Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZMYM4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599744
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599744 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599744 Supplier Invitrogen™ Supplier No. PA599744
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ZNF262.

Zinc-finger proteins contain DNA-binding domains characterized by the unique role of zinc and have a wide variety of functions such as transcriptional activation or repression. The protein folding and the DNA binding ability are governed by the coordination of a zinc ion. It has been found to be overexpressed in human lung adenocarcinomas and squamous cell carcinomas, and the overexpression of a fragment of the 3'UTR of the ZMYM4 mRNA termed Cell Death Inhibiting RNA (CDIR) protects HeLa cells from IFN-gamma-induced apoptosis, suggesting that ZMYM4 may play a role in tumorigenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZMYM4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ZMYM4
Gene Accession No. A2A791, Q5VZL5
Gene Alias 6330503C17Rik; AI480785; AW493829; CDIR; cell death inhibiting RNA; D630001M21; Kiaa0425; mKIAA0425; MYM; MYM type 4; Zfp262; zinc finger MYM-type containing 4; zinc finger MYM-type protein 4; Zinc finger protein 262; zinc finger, MYM-type 4; Zmym4; ZNF198L3; ZNF262
Gene Symbols ZMYM4
Host Species Rabbit
Immunogen A synthesized peptide derived from human ZMYM4(Accession Q5VZL5), corresponding to amino acid residues S804-C854.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 313598, 67785, 9202
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.