Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZMYND10 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA521864

Catalog No. PIPA521864


Only null left
Add to Cart

Description

Description

Recommended positive controls: 293T, A431, H1299, Raji. Predicted reactivity: Mouse (100%), Rat (100%), Chimpanzee (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The ZMYND10 gene encodes a protein that contains a MYND-type zinc finger domain, playing a crucial role in the assembly and function of cilia. Specifically, ZMYND10 is essential for the proper axonemal assembly of both inner and outer dynein arms, which are critical components of motile cilia. Mutations in ZMYND10 are associated with primary ciliary dyskinesia (PCD), a genetic disorder characterized by defective ciliary motility leading to recurrent respiratory infections and male infertility. In model organisms like mice and Drosophila, ZMYND10 expression is restricted to tissues and cells with motile cilia, such as chordotonal sensory neurons and sperm cells. Additionally, ZMYND10 has been identified as an epigenetically regulated tumor suppressor in various cancers, with its downregulation affecting processes like cell proliferation and tumor progression. The gene's dual role in ciliary function and tumorigenesis underlines its importance in both normal cellular physiology and disease pathology.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZMYND10
Polyclonal
Unconjugated
Zmynd10
Blu; CILD22; FLU; LUCA12.4; Protein BLu; zinc finger MYND domain-containing protein 10; zinc finger MYND-type containing 10; zinc finger, MYND domain containing 10; zinc finger, MYND domain-containing 10; zinc finger, MYND-type containing 10; Zmynd10
Rabbit
Antigen affinity chromatography
RUO
51364
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
O75800
Zmynd10
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human BLU. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.