Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZMYND11 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP286908
Description
ZMYND11 Polyclonal specifically detects ZMYND11 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ZMYND11 | |
Polyclonal | |
Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin | |
adenovirus 5 E1A binding protein, Adenovirus 5 E1A-binding protein, bone morphogenetic protein receptor-associated molecule 1, BRAM1, MGC111056, MYND domain containing 11, Protein BS69, zinc finger MYND domain-containing protein 11, zinc finger, MYND-type containing 11 | |
The immunogen is a synthetic peptide directed towards the middle region of human ZMYND11 | |
100 μL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS, 2% Sucrose | |
Rabbit | |
Protein A purified | |
RUO | |
10771 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction