Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZMYND11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540959
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540959 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540959 Supplier Invitrogen™ Supplier No. PA540959
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SMGWKKACDE LELHQRFLRE GRFWKSKNED RGEEEAESSI SSTSNEQLKV Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

The protein encoded by this gene was first identified by its ability to bind the adenovirus E1A protein. The protein localizes to the nucleus. It functions as a transcriptional repressor, and expression of E1A inhibits this repression. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZMYND11
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZMYND11
Gene Accession No. Q15326
Gene Alias 2210402G22Rik; adenovirus 5 E1A-binding protein; Bone morphogenetic protein receptor-associated molecule 1; BRAM1; BS69; MGC111056; MRD30; Protein BS69; RP11-486H9.1; Unknown (protein for MGC:137432); zinc finger MYND domain-containing protein 11; Zinc finger MYND domain-containing protein 11-like protein; zinc finger MYND-type containing 11; zinc finger, MYND domain containing 11; zinc finger, MYND-type containing 11; ZMYND11
Gene Symbols ZMYND11
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human ZMYND11.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10771
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.