Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZMYND11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA527899
Description
Recommended positive controls: BS69 tansfected 293T cell. Predicted reactivity: Mouse (97%), Rat (97%), Rhesus Monkey (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene was first identified by its ability to bind the adenovirus E1A protein. The protein localizes to the nucleus. It functions as a transcriptional repressor, and expression of E1A inhibits this repression. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
ZMYND11 | |
Polyclonal | |
Unconjugated | |
ZMYND11 | |
2210402G22Rik; adenovirus 5 E1A-binding protein; Bone morphogenetic protein receptor-associated molecule 1; BRAM1; BS69; MGC111056; MRD30; Protein BS69; RP11-486H9.1; Unknown (protein for MGC:137432); zinc finger MYND domain-containing protein 11; Zinc finger MYND domain-containing protein 11-like protein; zinc finger MYND-type containing 11; zinc finger, MYND domain containing 11; zinc finger, MYND-type containing 11; ZMYND11 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10771 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q15326 | |
ZMYND11 | |
Recombinant fragment corresponding to a region within amino acids 1 and 335 of Human BS69. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction