Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZMYND17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543561
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543561 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543561 Supplier Invitrogen™ Supplier No. PA543561
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SAHRGLYHDF WEEQVETGQT HHPDLVAAFH PGFHSSPDLM EAWLPTLLLL Sequence homology: Cow: 79%; Dog: 86%; Guinea Pig: 79%; Horse: 79%; Human: 100%; Rabbit: 79%.

ZMYND17 contains 1 MYND-type zinc finger. The exact function of ZMYND17 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZMYND17
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MSS51
Gene Accession No. Q4VC12
Gene Alias Mss51; MSS51 mitochondrial translational activator; MSS51 mitochondrial translational activator homolog; putative protein MSS51 homolog, mitochondrial; Zinc finger MYND domain-containing protein 17; zinc finger, MYND domain containing 17; zinc finger, MYND-type containing 17; ZMYND17
Gene Symbols MSS51
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human ZMYND17 (aa 326-375).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 118490
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.