Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZN438 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558951
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558951 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558951 Supplier Invitrogen™ Supplier No. PA558951
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PEKVHSSSEE SHACPRLKRQ LHLHQNGVEM LMENEGPQSG TNKPRETCQG PECPGLHTFL LWSHSGFNCL LCAEMLGRKE DLLHHWKHQH NCEDPSKLWA I Highest antigen sequence identity to the following orthologs: Mouse - 54%, Rat - 52%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZN438
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF438
Gene Accession No. Q7Z4V0
Gene Alias bA330O11.1; Zinc finger protein 438; ZNF438
Gene Symbols ZNF438
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZN438. Recombinant protein control fragment (Product #RP-95750).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 220929
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.