Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF692 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563704
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563704 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563704 Supplier Invitrogen™ Supplier No. PA563704
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GLVWECSAGH TFSWGPSLSP TPSEAPKPAS LPHTTRRSWC SEATSGQELA DLESEHDERT QEARLPRRVG PPPETFP Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 71%.

ZNF692, a zinc finger protein, is involved in several cellular processes including nucleolar organization and ribosome biogenesis. It interacts with nucleophosmin (NPM1), regulating nucleolar morphology by modifying NPM1's self-assembly properties and influencing protein synthesis. Knockdown of ZNF692 causes nucleolar redistribution of NPM1 and results in a decreased rate of protein synthesis. Additionally, ZNF692 plays a role in 40S ribosomal subunit maturation by forming a hub with the exosome complex and ribosome biogenesis factors. This gene is upregulated in certain cancers such as clear cell renal cell carcinoma (ccRCC), where its expression is associated with increased cell proliferation, migration, and poor overall survival.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF692
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF692
Gene Accession No. Q9BU19
Gene Alias AICAR responsive element binding protein; AREBP; Zfp692; zinc finger protein 692; ZNF692
Gene Symbols ZNF692
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF692. Recombinant protein control fragment (Product #RP-103352).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55657
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.