Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNDR1 Antibody (OTI1G2), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274930G
Description
ZNDR1 Monoclonal specifically detects ZNDR1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
ZNDR1 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ZNRD1 | |
Full length recombinant protein of human ZNDR1 (NP_740753) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI1G2 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
DNA-directed RNA polymerase I subunit RPA12, HTEX-6, hZR14, MGC13376, RNA polymerase I small specific subunit Rpa12, Rpa12, tctex-6, TEX6, transcription-associated zinc ribbon protein, zinc ribbon domain containing 1, zinc ribbon domain containing, 1, Zinc ribbon domain-containing protein 1 | |
Mouse | |
Affinity Purified | |
DNA replication Transcription Translation and Splicing | |
30834.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction