Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF10 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | ZNF10 |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1 - 2 ug/ml, Immunohistochemistry 2-4 ug/ml, Immunocytochemistry/Immunofluorescence 5 - 10 ug/ml, Immunohistochemistry-Paraffin 2-4 ug/ml |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Conjugate | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Conjugate | Quantity | Price | Quantity & Availability | ||||
NBP259679SS
![]() |
Novus Biologicals
NBP259679SS |
Unconjugated | 0.025 mg |
Each for $206.50
|
|
||||
NBP259679
![]() |
Novus Biologicals
NBP259679 |
Unconjugated | 0.1 mg |
Each for $499.50
|
|
||||
Description
ZNF10 Polyclonal specifically detects ZNF10 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
ZNF10 | |
Western Blot 1 - 2 ug/ml, Immunohistochemistry 2-4 ug/ml, Immunocytochemistry/Immunofluorescence 5 - 10 ug/ml, Immunohistochemistry-Paraffin 2-4 ug/ml | |
Polyclonal | |
Rabbit | |
Transcription Factors and Regulators | |
PBS with 0.02% Sodium Azide | |
7556 | |
Synthetic peptide made to an internal sequence of the human Znf10 protein (between amino acids 200-250) [UniProt P21506]. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1.0 mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
KOX1zinc finger protein 10 (KOX 1), zinc finger protein 10, Zinc finger protein KOX1 | |
ZNF10 | |
IgG | |
Affinity Purified |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title