Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF10 Antibody (CL9744), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP288939
Description
ZNF10 Monoclonal specifically detects ZNF10 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
ZNF10 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.2, containing 40% glycerol | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunofluorescence | |
CL9744 | |
Western Blot 1 ug/ml, Immunocytochemistry/Immunofluorescence 2-10 ug/ml | |
KOX1zinc finger protein 10 (KOX 1), zinc finger protein 10, Zinc finger protein KOX1 | |
This antibody was developed against a synthetic peptide corresponding to amino acids: RDQLDKYQENPERHLRQV | |
100 μL | |
Transcription Factors and Regulators | |
7556 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction