Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548868
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548868 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548868 Supplier Invitrogen™ Supplier No. PA548868
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF10 Polyclonal Antibody for Western Blot

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF10
Applications Western Blot
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ZNF10
Gene Accession No. P21506
Gene Alias KOX1; zinc finger protein 10; zinc finger protein 10 (KOX 1); zinc finger protein KOX1; ZNF10
Gene Symbols ZNF10
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 118-148 amino acids from the N-terminal region of human ZNF10.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7556
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.