Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF100 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297820MFV610
Description
ZNF100 Polyclonal antibody specifically detects ZNF100 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
ZNF100 | |
Polyclonal | |
Western Blot, Immunohistochemistry-Paraffin | |
FLJ44587, zinc finger protein 100, zinc finger protein 100 (Y1) | |
Produced in rabbits immunized with E. coli-derived Human ZNF100 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
163227 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction