Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF101 Monoclonal Antibody (2D5)
GREENER_CHOICE

Catalog No. PIMA522892
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522892 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522892 Supplier Invitrogen™ Supplier No. MA522892
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: LRHSFLDRHM RAHAGHKRSE CGGEWRETPR KQKQHGKASI SPSSGARRTV TPTRKRPYEC KVCGKAFNSP NLFQIHQRTH TGKRSYKCRE.

Zinc finger proteins (ZNFs), such as ZNF101, bind nucleic acids and perform many key functions, the most important of which is regulating transcription (summary by Bellefroid et al., 1993 ). See ZNF91 for general information on ZNFs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF101
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 2D5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene ZNF101
Gene Accession No. Q8IZC7
Gene Alias DKFZp570I0164; HZF12; MGC149565; MGC149566; zinc finger protein 101; zinc finger protein 101 (Y2); zinc finger protein 12; zinc finger protein HZF12; ZNF101
Gene Symbols ZNF101
Host Species Mouse
Immunogen ZNF101 (NP_149981.2, 112 a.a. ~ 201 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94039
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.