Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF101 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110595
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110595 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110595 Supplier Invitrogen™ Supplier No. PA5110595
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MRAHAGHKRS ECGGEWRETP RKQKQHGKAS ISPSSGARRT VTPTRKRPYE CKVCGKAFNS PNLFQIHQRT HTGKRSYKCR EIVRAFTVSS FFRKHGKMHT GEKRYECKYC GKPIDYPSLF QIHVRTHTGE KPYKCKQCGK AFISAGYLRT HEIRSHALEK SHQCQECGKK LSCSSSLHRH ERTHSGGKLY ECQKCAKVFR CPTSLQAHER AHTGERPYEC NKCGKTFNYP SCFRRHKKTH SGEKPYECTR CGKAFGWCSS LRRHEMTHTG EKPFDCKQCG KVFTFSNYLR LHERTHLAGR SQCFGRRQGD HLSPGV.

Zinc finger proteins (ZNFs), such as ZNF101, bind nucleic acids and perform many key functions, the most important of which is regulating transcription (summary by Bellefroid et al., 1993 ). See ZNF91 for general information on ZNFs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF101
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ZNF101
Gene Accession No. Q8IZC7
Gene Alias DKFZp570I0164; HZF12; MGC149565; MGC149566; zinc finger protein 101; zinc finger protein 101 (Y2); zinc finger protein 12; zinc finger protein HZF12; ZNF101
Gene Symbols ZNF101
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 121-436 of human ZNF101 (NP_149981.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94039
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.