Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF114 Antibody (OTI6F4), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274931AF405
Description
ZNF114 Monoclonal specifically detects ZNF114 in Human samples. It is validated for Western Blot.Specifications
ZNF114 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ZNF114 | |
Full length recombinant protein of human ZNF114 (NP_705836) produced in E.coli. | |
0.1 mL | |
163071.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI6F4 | |
Western Blot | |
MGC149700, MGC17986, zinc finger protein 114, zinc finger protein 114 (Y18) | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction