Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF114 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555204
Description
Immunogen sequence: LPKRTFPEAN RVCLTSISSQ HSTLREDWRC PKTEEPHRQG VNNVKPPAVA PEKDESPVSI CEDHEMRNHS KPTC Highest antigen sequence identity to the following orthologs: Mouse - 26%, Rat - 28%.
ZNF114 may be involved in transcriptional regulation.
Specifications
| ZNF114 | |
| Polyclonal | |
| Unconjugated | |
| ZNF114 | |
| Gm1957; Zfp110; Zfp114; Zinc finger protein 114; zinc finger protein 114 (Y18); ZNF114 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 163071 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NC26 | |
| ZNF114 | |
| Recombinant protein corresponding to Human ZNF114. Recombinant protein control fragment (Product #RP-93621). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction