Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF114 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556187
Description
Immunogen sequence: DSIRQCILTR DSSIFKYNPV LNDSQKTHEN NEDDGVLGWN IQWVPCGRKT ELKSSTWTGS QNTVHHIRDE IDTG Highest antigen sequence identity to the following orthologs: Mouse - 26%, Rat - 26%.
ZNF114 may be involved in transcriptional regulation.
Specifications
ZNF114 | |
Polyclonal | |
Unconjugated | |
ZNF114 | |
Gm1957; Zfp110; Zfp114; Zinc finger protein 114; zinc finger protein 114 (Y18); ZNF114 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
163071 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NC26 | |
ZNF114 | |
Recombinant protein corresponding to Human ZNF114. Recombinant protein control fragment (Product #RP-93620). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction