Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF117 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543074
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543074 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543074 Supplier Invitrogen™ Supplier No. PA543074
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: EIPYKCEKCV RAFNQASKLT EHKLIHTGEK RYECEECGKA FNRSSKLTEH Sequence homology: Dog: 91%; Human: 100%.

ZNF117 belongs to the krueppel C2H2-type zinc-finger protein family and may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF117
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF117
Gene Accession No. Q03924
Gene Alias HPF9; h-PLK; Krueppel-related zinc finger protein; provirus-linked krueppel; Zinc finger protein 117; zinc finger protein HPF9; ZNF117
Gene Symbols ZNF117
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human ZNF117.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51351
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.