Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541149
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541149 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541149 Supplier Invitrogen™ Supplier No. PA541149
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: FLLQSYPDEV WQTDDLIERI QEEENKPSRQ TVFIETLIEE RGNVPGKTFD Sequence homology: Human: 100%.

Two members of the human zinc finger Kruppel family, ZNF 12 (KOX 3) and ZNF 26 (KOX 20), have been localized by somatic cell hybrid analysis and in situ chromosomal hybridization. The presence of individual human zinc finger genes in mouse-human hybrid DNAs was correlated with the presence of specific human chromosomes or regions of chromosomes in the corresponding cell hybrids. Analysis of such mouse-human hybrid DNAs assigned the ZNF 12 (KOX 3) gene to chromosome region 7p.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF12
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Znf12
Gene Accession No. P17014
Gene Alias C530015C18; GIOT3; GIOT-3; gonadotropin inducible transcription repressor 3; gonadotropin-inducible ovary transcription repressor 3; HZF11; KOX3; Krox-7; Zfp12; Zfp-12; zinc finger protein 11; zinc finger protein 12; Zinc finger protein 325; zinc finger protein KOX3; Znf12; ZNF325
Gene Symbols Znf12
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ZNF12 (aa 73-122).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7559
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.