Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF131 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543066
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543066 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543066 Supplier Invitrogen™ Supplier No. PA543066
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: EFLQMLEAIK ALEVRNKENS APLEENTTGK NEAKKRKIAE TSNVITESLP Sequence homology: Cow: 93%; Dog: 93%; Horse: 86%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.

May be involved in transcriptional regulation. Plays a role during development and organogenesis as well as in the function of the adult central nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF131
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF131
Gene Accession No. P52739
Gene Alias 2610109I01Rik; AW558871; pHZ-10; ZBTB35; Zfp131; zinc finger and BTB domain containing 35; zinc finger protein 131; zinc finger protein 131 (clone pHZ-10); Zinc finger protein 131-like protein; ZNF131
Gene Symbols ZNF131
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ZNF131 (aa 110-159).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7690
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.