Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF134 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541098
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541098 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541098 Supplier Invitrogen™ Supplier No. PA541098
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SEKPFTCKEE QKNFQATLGG CQQKAIHSKR KTHRSTESGD AFHGEQMHYK Sequence homology: Human: 100%; Pig: 86%.

Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Krueppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. Zinc-finger protein 134 (ZNF134) is a 348 amino acid member of the Krueppel C2H2-type zinc-finger protein family. ZNF134 localizes to the nucleus and contains nine C2H2-type zinc fingers through which it is thought to be involved in DNA-binding and transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF134
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF134
Gene Accession No. P52741
Gene Alias pHZ-15; zinc finger protein 134; zinc finger protein 134 (clone pHZ-15); ZNF134
Gene Symbols ZNF134
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human ZNF134.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7693
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.