Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF134 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA590662
Description
Positive Samples: Rat liver; Cellular Location: Nucleus Immunogen sequence: HGLKLHTCGA CGRQFWFSAN LHQYQKCYSI EQPLRRDKSE ASIVKNCTVS KEPHPSEKPF TCKEEQKNFQ ATLGGCQQKA IHSKRKTHRS TESGDAFH.
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Krueppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. Zinc-finger protein 134 (ZNF134) is a 348 amino acid member of the Krueppel C2H2-type zinc-finger protein family. ZNF134 localizes to the nucleus and contains nine C2H2-type zinc fingers through which it is thought to be involved in DNA-binding and transcriptional regulation.
Specifications
ZNF134 | |
Polyclonal | |
Unconjugated | |
ZNF134 | |
pHZ-15; zinc finger protein 134; zinc finger protein 134 (clone pHZ-15); ZNF134 | |
Rabbit | |
Affinity chromatography | |
RUO | |
7693 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.53 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P52741 | |
ZNF134 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 73-170 of human ZNF134 (NP_0034263). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction