Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF135 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552368
Description
Immunogen sequence: FLWDGLWYCR GEDTEGHWEW SCESLESLAV PVAFTPVKTP VLEQWQRNGF GENISLNPDL PHQPMTPERQ SPHTWGTRGK REKPDLNVLQ KTCVKEKPYK CQECGKAFSH SSALI Highest antigen sequence identity to the following orthologs: Mouse - 29%, Rat - 31%.
ZNF135 may be involved in transcriptional regulation.
Specifications
ZNF135 | |
Polyclonal | |
Unconjugated | |
ZNF135 | |
pHZ-17; pT3; Zinc finger protein 135; zinc finger protein 135 (clone pHZ-17); zinc finger protein 61; Zinc finger protein 78-like 1; zinc finger protein 78-like 1 (pT3); ZNF135; ZNF61; ZNF78L1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7694 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P52742 | |
ZNF135 | |
Recombinant protein corresponding to Human ZNF135. Recombinant protein control fragment (Product #RP-89430). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction