Learn More
Invitrogen™ ZNF136 Monoclonal Antibody (5E9)
Mouse Monoclonal Antibody
Supplier:  Invitrogen™ MA520271
Catalog No. PIMA520271
Only null left
Add to Cart
Description
Description
Peptide Sequence: GEKPYECKQC GKAFSYLNSF RTHEMIHTGE KPFECKRCGK AFRSSSSFRL HERTHTGQKP YHCKECGKAY SCRASFQRHM LTHAEDGPPY KCMWESL.
ZNF136 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 14 C2H2-type zinc fingers and 1 KRAB domain. ZNF136 may be involved in transcriptional regulation as a weak repressor when alone, or a potent one when fused with a heterologous protein containing a KRAB B-domain.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ZNF136
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
P52737
ZNF136
ZNF136 (NP_003428, 444 a.a. ~ 540 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG2a κ
ELISA, Western Blot, Immunocytochemistry
5E9
Unconjugated
ZNF136
pHZ-20; zinc finger protein 136; ZNF136
Mouse
Affinity chromatography
RUO
7695
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.