Learn More
Invitrogen™ ZNF136 Polyclonal Antibody

Description
Immunogen sequence: ECGKAYSCRAS FQRHMLTHAE DGPPYKCMWE Highest antigen sequence identity to the following orthologs - mouse 52%, rat 52%.
Specifications
Specifications
| Antigen | ZNF136 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ZNF136 |
| Gene Accession No. | P52737 |
| Gene Alias | pHZ-20; zinc finger protein 136; ZNF136 |
| Gene Symbols | ZNF136 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.