Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF138 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569490
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569490 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569490 Supplier Invitrogen™ Supplier No. PA569490
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 83%; Human: 100%; Mouse: 85%; Rat: 85%; Zebrafish: 79%.

May be involved in transcriptional regulation as a repressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF138
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF138
Gene Accession No. P52744
Gene Alias pHZ-32; Zinc finger protein 138; zinc finger protein 138 (clone pHZ-32); ZNF138
Gene Symbols ZNF138
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human ZNF138.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7697
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.