Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543693
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543693 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543693 Supplier Invitrogen™ Supplier No. PA543693
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: IYYQPFQRHE RTHAGQKPYE CKQCGKTFIY YQSFQKHAHT GKKPYECKQC Sequence homology: Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

ZNF14 contains a zinc finger and a Kruppel-associated box (KRAB) domain. KRAB domain is known to be involved in the transcriptional repression of a number of zinc finger proteins.The protein encoded by this gene contains a zinc finger and a Kruppel-associated box (KRAB) domain. KRAB domain is known to be involved in the transcriptional repression of a number of zinc finger proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF14
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF14
Gene Accession No. P17017
Gene Alias GIOT4; GIOT-4; gonadotropin inducible transcription repressor-4; gonadotropin-inducible ovary transcription repressor 4; KOX6; Zinc finger protein 14; zinc finger protein 14 (KOX 6); zinc finger protein KOX6; ZNF14
Gene Symbols ZNF14
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ZNF14 (aa 179-228).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7561
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.