Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF140 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561714
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561714 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561714 Supplier Invitrogen™ Supplier No. PA561714
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GKREVKRDLF SVSESSGEIK DFSPKNVIYD DSSQYLIMER ILSQGPVYSS FKGGWKCKDH TEMLQENQGC IRKVTVSHQE AL Highest antigen sequence identity to the following orthologs: Mouse - 28%, Rat - 26%.

The ZNF140 gene encodes a zinc finger protein that contains KRAB (Kruppel-associated box) domains, which are known to function as transcriptional repressors. These domains are located at the N-terminus of the protein and are involved in mediating protein-protein interactions, critical for gene silencing. ZNF140 is part of a large family of zinc finger proteins, which play significant roles in regulating gene expression by binding to specific DNA sequences. The KRAB domains of ZNF140 enable it to interact with other transcriptional repressors and recruit corepressor complexes, leading to chromatin remodeling and transcriptional repression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF140
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF140
Gene Accession No. P52738
Gene Alias pHZ-39; Zinc finger protein 140; ZNF140
Gene Symbols ZNF140
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF140. Recombinant protein control fragment (Product #RP-100882).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7699
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.