Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF143 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572658
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572658 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572658 Supplier Invitrogen™ Supplier No. PA572658
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF143 Polyclonal Antibody for Western Blot

Transcriptional activator. Activates the gene for selenocysteine tRNA (tRNAsec). Binds to the SPH motif of small nuclear RNA (snRNA) gene promoters. Participates in efficient U6 RNA polymerase III transcription via its interaction with CHD8.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF143
Applications Western Blot
Classification Polyclonal
Concentration 0.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene znf143b
Gene Accession No. Q1LYE3
Gene Alias AA959806; AU015869; D7Ertd805e; hStaf; KRAB14; mStaf; pHZ-1; SBF; selenocysteine tRNA activating factor; selenocysteine tRNA gene transcription activating factor; selenocysteine tRNA gene transcription-activating factor; si:dkey-8l13.2; SPH-binding factor; staf; transcriptional activator Staf; Zfp143; zfp-143; Zfp79; Zfp80-rs1; zgc:66276; zinc finger protein 143; zinc finger protein 143b; zinc finger protein 80, related sequence 1; znf143; znf143b
Gene Symbols znf143b
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 564-596 amino acids of DANRE znf143.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 393954
Target Species Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.