Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF143 Polyclonal Antibody

Supplier: Thermo Scientific PA5118371
This item is not returnable.
View return policy
Description
Specifications
| ZNF143 | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| P52747 | |
| ZNF143 | |
| E. coli-derived Human ZNF143 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| ZNF143 | |
| AA959806; AU015869; D7Ertd805e; hStaf; KRAB14; mStaf; pHZ-1; SBF; selenocysteine tRNA activating factor; selenocysteine tRNA gene transcription activating factor; selenocysteine tRNA gene transcription-activating factor; si:dkey-8l13.2; SPH-binding factor; staf; transcriptional activator Staf; Zfp143; zfp-143; Zfp79; Zfp80-rs1; zgc:66276; zinc finger protein 143; zinc finger protein 143b; zinc finger protein 80, related sequence 1; znf143; znf143b | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 7702 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction