Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF143 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120557
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120557 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120557 Supplier Invitrogen™ Supplier No. PA5120557
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: U-87MG, HepG2, Mouse spleen, Mouse liver, Rat testis, Rat liver. Immunogen sequence: RTAHNDTEPI EEEQEAFFEP PPGQGEDVLK GSQITYVTGV EGDDVVSTQV ATVTQSGLSQ QVTLISQDGT QHVNISQADM QAIGNTITMV TQDGTPITVP AHDAVISSAG THSVAMVTAE GTEGEQVAIV AQDLAAFHTA SSEMGHQQHS HHLVTTETRP LTLVATSNGT QIAVQLGEQP SLEEAIRIAS RIQQGETPGL DD.

Transcriptional activator. Activates the gene for selenocysteine tRNA (tRNAsec). Binds to the SPH motif of small nuclear RNA (snRNA) gene promoters. Participates in efficient U6 RNA polymerase III transcription via its interaction with CHD8.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF143
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.26 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ZNF143
Gene Accession No. O70230, P52747, Q5XIU2
Gene Alias AA959806; AU015869; D7Ertd805e; hStaf; KRAB14; mStaf; pHZ-1; SBF; selenocysteine tRNA activating factor; selenocysteine tRNA gene transcription activating factor; selenocysteine tRNA gene transcription-activating factor; si:dkey-8l13.2; SPH-binding factor; staf; transcriptional activator Staf; Zfp143; zfp-143; Zfp79; Zfp80-rs1; zgc:66276; zinc finger protein 143; zinc finger protein 143b; zinc finger protein 80, related sequence 1; znf143; znf143b
Gene Symbols Zfp143, ZNF143
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 437-638 of human ZNF143 (NP_003433.3).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20841, 361627, 7702
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.