Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF146 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297822APC
Description
ZNF146 Polyclonal antibody specifically detects ZNF146 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
ZNF146 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
MGC125660, MGC125661, only zinc finger protein, OZF, zinc finger protein 146, zinc finger protein OZF | |
Produced in rabbits immunized with E. coli-derived Human ZNF146 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
7705 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction