Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF146 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551908
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551908 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551908 Supplier Invitrogen™ Supplier No. PA551908
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSQQRIYSGE NPFACKVCGK VFSHKSNLTE HEHFHTREKP FECNECGKAF SQKQYVIKHQ NTHTGEKLFE CNECGKSFSQ KENLLTHQKI HTGEKPFECK DCGKAFIQKS NLIR Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 94%.

ZNF146 (zinc finger protein 146), also known as OZF (only zinc finger protein), is a 292 amino acid nuclear protein that binds DNA and belongs to the Kruppel C2H2-type zinc-finger protein family. ZNF146 interacts with UBC9 (ubiquitin carrier protein 9) and RAP1 (repressor/activator protein 1 homolog), and is highly expressed in multiple pancreatic and colorectal cancers. Lower levels of expression have been observed in kidney, lung, placenta and brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF146
Applications ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF146
Gene Accession No. Q15072
Gene Alias Only zinc finger protein; OZF; Zinc finger protein 146; zinc finger protein OZF; ZNF146
Gene Symbols LOC102643033, ZNF146
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF146. Recombinant protein control fragment (Product #RP-100741).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102643033, 103691098, 7705
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.