Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF148 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171872UV
Description
ZNF148 Polyclonal antibody specifically detects ZNF148 in Human, Mouse samples. It is validated for Western BlotSpecifications
ZNF148 | |
Polyclonal | |
Western Blot | |
BERF-1, BFCOL1, CACCC box-binding protein, CLL-associated antigen KW-10, HT-BETA, pHZ-52, Transcription factor ZBP-89, ZBP89, ZBP-89, ZFP148zinc finger protein 148 (pHZ-52), Zinc finger DNA-binding protein 89, zinc finger protein 148 | |
The immunogen for this product maps to a region between residue 744 and 794 of human Zinc Finger DNA-Binding Protein 89 using the numbering given in entry NP_068799.2 (GeneID 7707). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
7707 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction