Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF148 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$377.03 - $666.47
Specifications
| Antigen | ZNF148 |
|---|---|
| Dilution | Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20-1:50 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
ZNF148 Polyclonal specifically detects ZNF148 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| ZNF148 | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 7707 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:MQSSRTMVVMGGVSGQSTVSGELQDSVLQDRSMPHQEILAADEVLQESEMRQQDMISHDELMVHEETVKNDEEQMETHERLPQGLQYALNVPISVKQEITFTDVSEQLMRDK | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20-1:50 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| BERF-1, BFCOL1, CACCC box-binding protein, CLL-associated antigen KW-10, HT-BETA, pHZ-52, Transcription factor ZBP-89, ZBP89, ZBP-89, ZFP148zinc finger protein 148 (pHZ-52), Zinc finger DNA-binding protein 89, zinc finger protein 148 | |
| ZNF148 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title