Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF148 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551620
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551620 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551620 Supplier Invitrogen™ Supplier No. PA551620
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MQSSRTMVVM GGVSGQSTVS GELQDSVLQD RSMPHQEILA ADEVLQESEM RQQDMISHDE LMVHEETVKN DEEQMETHER LPQGLQYALN VPISVKQEIT FTDVSEQLMR DK Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.

ZNF148 is involved in transcriptional regulation. ZNF148 represses the transcription of a number of genes including gastrin, stromelysin and enolase. ZNF148 binds to the G-rich box in the enhancer region of these genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF148
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF148
Gene Accession No. Q9UQR1
Gene Alias 2210405J08Rik; AI480666; AW045217; BERF-1; Beta enolase repressor factor 1; BFCOL1; CACCC box-binding protein; CLL-associated antigen KW-10; fb06h05; G-rich box-binding protein; HT-BETA; pHZ-52; si:dkey-101k6.6; transcription factor; transcription factor BFCOL1; Transcription factor ZBP-89; wu:fb06h05; Zbp89; ZBP-89; Zfp148; zfp149; zgc:165342; zinc finger binding protein 89; zinc finger DNA binding protein 89; zinc finger DNA-binding protein 89; Zinc finger protein 148; znf148
Gene Symbols ZNF148
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF148. Recombinant protein control fragment (Product #RP-100702).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7707
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.