Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF148 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569021
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569021 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569021 Supplier Invitrogen™ Supplier No. PA569021
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 93%; Horse: 100%; Human: 100%; Mouse: 86%; Pig: 93%; Rat: 93%.

ZNF148 is involved in transcriptional regulation. ZNF148 represses the transcription of a number of genes including gastrin, stromelysin and enolase. ZNF148 binds to the G-rich box in the enhancer region of these genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF148
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Zfp148
Gene Accession No. Q61624
Gene Alias 2210405J08Rik; AI480666; AW045217; BERF-1; Beta enolase repressor factor 1; BFCOL1; CACCC box-binding protein; CLL-associated antigen KW-10; fb06h05; G-rich box-binding protein; HT-BETA; pHZ-52; si:dkey-101k6.6; transcription factor; transcription factor BFCOL1; Transcription factor ZBP-89; wu:fb06h05; Zbp89; ZBP-89; Zfp148; zfp149; zgc:165342; zinc finger binding protein 89; zinc finger DNA binding protein 89; zinc finger DNA-binding protein 89; Zinc finger protein 148; znf148
Gene Symbols Zfp148
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of mouse ZNF148.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22661
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.