Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF154 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5140032
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: LAKLGFLHQQ AAHTGEQSNS KSDGGAISHR GKTHYNCGEH TKAFSGKHTL VQQQRTLTTE RCY.
ZNF154 may be involved in transcriptional regulation.
Specifications
ZNF154 | |
Polyclonal | |
Unconjugated | |
ZNF154 | |
KIAA2003; pHZ-92; Zinc finger protein 154; ZNF154 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7710 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q13106 | |
ZNF154 | |
Recombinant protein corresponding to Human ZNF154. Recombinant protein control fragment (Product #RP-109549). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction