Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF157 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP132389
Description
ZNF157 Polyclonal specifically detects ZNF157 in Human samples. It is validated for Western Blot.Specifications
ZNF157 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
ZNF157 | |
Recombinant protein encompassing a sequence within the center region of human ZNF157. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
7712 | |
Human | |
IgG |
Western Blot | |
0.45mg/mL | |
Western Blot 1:500-1:3000 | |
HZF22, zinc finger protein 157, zinc finger protein 157 (HZF22), zinc finger protein 22, Zinc finger protein HZF22 | |
Rabbit | |
58 kDa | |
0.1 mL | |
Primary | |
Tested in human. Not yet tested in any other species. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction