Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF157 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551893
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551893 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551893 Supplier Invitrogen™ Supplier No. PA551893
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YSNLASVGLC VAKPEMIFKL ERGEELWILE EESSGHGYSG SLSLLCGNGS VGDNALRHDN DLLHHQKIQT LDQNVEYNGC RKAFHEKTGF VRRKRTPRGD KNFECHECGK AYCRKSNLVE HLRIHTGER Highest antigen sequence identity to the following orthologs: Mouse - 33%, Rat - 33%.

This gene product is a likely zinc finger family transcription factor. It contains KRAB-A and KRAB-B domains that act as transcriptional repressors in related proteins, and multiple zinc finger DNA binding motifs and finger linking regions characteristic of the Kruppel family. This gene is part of a gene cluster on chromosome Xp11.23.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF157
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF157
Gene Accession No. P51786
Gene Alias HZF22; Zinc finger protein 157; zinc finger protein 22; zinc finger protein HZF22; ZNF157
Gene Symbols ZNF157
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF157. Recombinant protein control fragment (Product #RP-92212).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7712
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.