Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF157 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589753
Description
Immunogen sequence: VAKPEMIFKL ERGEELWILE EESSGHGYSG SLSLLCGNGS VGDNALRHDN DLLHHQKIQT LDQNVEYNGC RKAFHEKTGF VRRKRTPRGD KNFECH; Positive Samples: U-87MG; Cellular Location: Nucleus.
This gene product is a likely zinc finger family transcription factor. It contains KRAB-A and KRAB-B domains that act as transcriptional repressors in related proteins, and multiple zinc finger DNA binding motifs and finger linking regions characteristic of the Kruppel family. This gene is part of a gene cluster on chromosome Xp11.23.
Specifications
ZNF157 | |
Polyclonal | |
Unconjugated | |
ZNF157 | |
HZF22; Zinc finger protein 157; zinc finger protein 22; zinc finger protein HZF22; ZNF157 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
7712 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.07 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
P51786 | |
ZNF157 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 70-165 of human ZNF157 (NP_0034372). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction