Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF16 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557398
Description
Immunogen sequence: LSVPGPSPWT PAAQARVRDA PAVTHPGSAA CGTPCCSDTE LEAICPHYQQ PDCDTRTEDK EFLHKEDIHE DLESQAEISE NYA Highest antigen sequence identity to the following orthologs: Mouse - 28%, Rat - 28%.
ZNF16 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 17 C2H2-type zinc fingers. ZNF16 may be involved in transcriptional regulation.The protein encoded by this gene contains a C2H2 type of zinc finger, and thus may function as a transcription factor. This gene is located in a region close to ZNF7/KOX4, a gene also encoding a zinc finger protein, on chromosome 8. Two alternatively spliced variants, encoding the same protein, have been identified.
Specifications
| ZNF16 | |
| Polyclonal | |
| Unconjugated | |
| ZNF16 | |
| HZF1; KOX9; Zinc finger protein 16; zinc finger protein KOX9; zinc finger protein Kruppel type 9; ZNF16 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7564 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P17020 | |
| ZNF16 | |
| Recombinant protein corresponding to Human ZNF16. Recombinant protein control fragment (Product #RP-95527). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction