Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF165 Antibody (OTI4C1), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274932AF532
Description
ZNF165 Monoclonal specifically detects ZNF165 in Human samples. It is validated for Western Blot.Specifications
ZNF165 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
ZNF165 | |
Recombinant protein fragment corresponding to amino acids 165-485 of human ZNF165 (NP_003438) produced in E.coli. | |
Primary | |
Human | |
IgG1 |
Western Blot | |
OTI4C1 | |
Western Blot | |
Cancer/testis antigen 53, CT53Zinc finger and SCAN domain-containing protein 7, zinc finger protein 165, ZSCAN7LD65ZPF165 | |
Mouse | |
0.1 mL | |
7718 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction