Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF165 Antibody (OTI4C1), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274932C
Description
ZNF165 Monoclonal specifically detects ZNF165 in Human samples. It is validated for Western Blot.Specifications
ZNF165 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ZNF165 | |
Recombinant protein fragment corresponding to amino acids 165-485 of human ZNF165 (NP_003438) produced in E.coli. | |
0.1 mL | |
7718.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI4C1 | |
Western Blot | |
Cancer/testis antigen 53, CT53Zinc finger and SCAN domain-containing protein 7, zinc finger protein 165, ZSCAN7LD65ZPF165 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction